Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-1
Entrez Gene ID
191436
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_ZK829.8
Chromosome Location
Chr.IV: 11963910 - 11965427 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069869.1 NP_502270.1 1 - 2 187 169

View Full Product Details