Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spt-5
Entrez Gene ID
178143
Gene Name
Transcription elongation factor SPT5
Gene Aliases
CELE_K08E4.1
Chromosome Location
Chr.IV: 12051491 - 12055746 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069882.6 NP_502283.1 2 - 3 137 128

View Full Product Details