Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-37
Entrez Gene ID
187161
Gene Name
protein-coding
Gene Aliases
CELE_K08E4.5
Chromosome Location
Chr.IV: 12068890 - 12074948 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307301.3 NP_001294230.1 6 - 7 1036 80
NM_069886.4 NP_502287.2 6 - 7 1197 80

View Full Product Details