Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aly-3
Entrez Gene ID
178157
Gene Name
FoP_duplication domain-containing protein
Gene Aliases
CELE_M18.7
Chromosome Location
Chr.IV: 12123289 - 12124809 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083228.5 NP_001076697.1 4 - 5 567 72
NR_003454.1 3 - 4 958 72

View Full Product Details