Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-18
Entrez Gene ID
189745
Gene Name
protein-coding
Gene Aliases
CELE_Y39C12A.3
Chromosome Location
Chr.IV: 12227717 - 12229779 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306971.3 NP_001293900.1 2 - 3 187 113
NM_001307201.4 NP_001294130.1 7 - 8 847 113
NR_131614.1 6 - 7 689 113

View Full Product Details