Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-16
Entrez Gene ID
189747
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_Y39C12A.5
Chromosome Location
Chr.IV: 12234085 - 12236111 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069922.3 NP_502323.2 3 - 4 300 75

View Full Product Details