Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-19
Entrez Gene ID
189748
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_Y39C12A.6
Chromosome Location
Chr.IV: 12237175 - 12238951 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069923.1 NP_502324.1 2 - 3 257 121

View Full Product Details