Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-15
Entrez Gene ID
189749
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_Y39C12A.7
Chromosome Location
Chr.IV: 12241296 - 12244806 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069924.6 NP_502325.4 4 - 5 490 162

View Full Product Details