Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pgp-1
Entrez Gene ID
178215
Gene Name
Multidrug resistance protein pgp-1
Gene Aliases
CELE_K08E7.9
Chromosome Location
Chr.IV: 12591695 - 12599169 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070012.5 NP_502413.1 13 - 14 3897 69

View Full Product Details