The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001383222.1 | 23 - 24 | 3291 | 74 | |
NM_070018.6 | NP_502419.2 | 18 - 19 | 2559 | 74 | |
GenBank mRNA | AB055112.1 | - | 20 - 21 | 2956 | 74 |