Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-31
Entrez Gene ID
178233
Gene Name
protein-coding
Gene Aliases
CELE_ZK897.1
Chromosome Location
Chr.IV: 12773862 - 12788354 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268734.3 NP_001255663.1 10 - 11 1607 109
NM_001268735.4 NP_001255664.1 9 - 10 1586 109
NM_001268736.3 NP_001255665.1 8 - 9 1550 109
NM_001268737.4 NP_001255666.1 10 - 11 1641 109
NM_001268738.3 NP_001255667.1 9 - 10 1610 109
NM_001268739.3 NP_001255668.1 1 - 2 242 109
NM_001268740.3 NP_001255669.1 1 - 2 242 109
NM_001306972.3 NP_001293901.1 11 - 12 1643 109
NM_001306973.3 NP_001293902.1 11 - 12 1643 109
NM_001306974.3 NP_001293903.1 10 - 11 1622 109
NM_001306975.3 NP_001293904.1 10 - 11 1622 109
NM_001306976.3 NP_001293905.1 9 - 10 1586 109
NM_001306977.3 NP_001293906.1 9 - 10 1586 109
NM_001306978.3 NP_001293907.1 10 - 11 1607 109
NM_001306979.3 NP_001293908.1 9 - 10 1586 109
NM_001306980.3 NP_001293909.1 8 - 9 1550 109
NM_001306981.3 NP_001293910.1 1 - 2 242 109

View Full Product Details