Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
amt-2
Entrez Gene ID
186047
Gene Name
Putative ammonium transporter 2
Gene Aliases
CELE_F49E11.3
Chromosome Location
Chr.IV: 13042391 - 13047043 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070095.4 NP_502496.3 7 - 8 1168 95

View Full Product Details