Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scl-6
Entrez Gene ID
183343
Gene Name
SCP domain-containing protein
Gene Aliases
CELE_C39E9.4
Chromosome Location
Chr.IV: 13068751 - 13069994 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070107.2 NP_502508.1 2 - 3 200 59

View Full Product Details