Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scl-7
Entrez Gene ID
183344
Gene Name
SCP domain-containing protein
Gene Aliases
CELE_C39E9.5
Chromosome Location
Chr.IV: 13070442 - 13071270 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070108.3 NP_502509.1 2 - 3 569 70

View Full Product Details