Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rfc-3
Entrez Gene ID
178259
Gene Name
RFC (DNA replication factor) family
Gene Aliases
CELE_C39E9.13
Chromosome Location
Chr.IV: 13095762 - 13097080 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070116.8 NP_502517.1 2 - 3 896 67

View Full Product Details