Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-14
Entrez Gene ID
184247
Gene Name
Zinc metalloproteinase nas-14
Gene Aliases
CELE_F09E8.6
Chromosome Location
Chr.IV: 13170541 - 13174144 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070132.4 NP_502533.2 2 - 3 370 164

View Full Product Details