Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lsm-3
Entrez Gene ID
178303
Gene Name
protein-coding
Gene Aliases
CELE_Y62E10A.12
Chromosome Location
Chr.IV: 13387203 - 13389141 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070178.6 NP_502579.1 1 - 2 128 114

View Full Product Details