The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307155.3 | NP_001294084.1 | 12 - 13 | 3510 | 77 |
NM_070212.4 | NP_502613.2 | 17 - 18 | 4317 | 77 |