Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-10
Entrez Gene ID
191800
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_Y45F10B.14
Chromosome Location
Chr.IV: 13575033 - 13576806 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070213.4 NP_502614.1 3 - 4 721 68

View Full Product Details