Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-11
Entrez Gene ID
189917
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_Y45F10B.6
Chromosome Location
Chr.IV: 13584946 - 13587288 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070217.2 NP_502618.2 2 - 3 444 141

View Full Product Details