Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-9
Entrez Gene ID
189920
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_Y45F10B.11
Chromosome Location
Chr.IV: 13588993 - 13590337 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070219.2 NP_502620.2 3 - 4 588 116

View Full Product Details