Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-22
Entrez Gene ID
178330
Gene Name
UDP-glucuronosyltransferase
Gene Aliases
CELE_C08F11.8
Chromosome Location
Chr.IV: 13639260 - 13644402 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070232.6 NP_502633.1 5 - 6 1048 75

View Full Product Details