Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nkcc-1
Entrez Gene ID
3565648
Gene Name
protein-coding
Gene Aliases
CELE_Y37A1C.1
Chromosome Location
Chr.IV: 14107720 - 14124056 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083255.3 NP_001076724.1 4 - 5 978 92
NM_001268861.4 NP_001255790.1 5 - 6 1042 92
NM_001268862.4 NP_001255791.1 4 - 5 950 92
NM_001276883.3 NP_001263812.1 4 - 5 949 92
NM_070303.2 NP_502704.1 4 - 5 978 92

View Full Product Details