Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nkcc-1
Entrez Gene ID
3565648
Gene Name
protein-coding
Gene Aliases
CELE_Y37A1C.1
Chromosome Location
Chr.IV: 14107720 - 14124056 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083255.3 NP_001076724.1 9 - 10 2620 82
NM_001268859.3 NP_001255788.1 5 - 6 1571 82
NM_001268860.3 NP_001255789.1 2 - 3 605 82
NM_001268861.4 NP_001255790.1 10 - 11 2684 82
NM_001268862.4 NP_001255791.1 9 - 10 2592 82
NM_001276883.3 NP_001263812.1 9 - 10 2591 82
NM_070303.2 NP_502704.1 9 - 10 2620 82

View Full Product Details