Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-38
Entrez Gene ID
178369
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_H12I19.5
Chromosome Location
Chr.IV: 14150331 - 14153361 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307022.2 NP_001293951.1 9 - 10 1589 101

View Full Product Details