Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sqrd-1
Entrez Gene ID
178374
Gene Name
protein-coding
Gene Aliases
CELE_F02H6.5
Chromosome Location
Chr.IV: 14219860 - 14224440 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268869.4 NP_001255798.1 3 - 4 622 103
NM_001268870.3 NP_001255799.1 3 - 4 633 103
NM_001268871.3 NP_001255800.1 2 - 3 277 103
NM_001307012.3 NP_001293941.1 2 - 3 277 103

View Full Product Details