Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arl-8
Entrez Gene ID
178405
Gene Name
ARF-Like
Gene Aliases
CELE_Y57G11C.13
Chromosome Location
Chr.IV: 14798149 - 14799229 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070390.9 NP_502791.1 1 - 2 157 105
GenBank mRNA AB185209.1 - 1 - 2 127 105

View Full Product Details