Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-47
Entrez Gene ID
189832
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_Y41E3.12
Chromosome Location
Chr.IV: 15049380 - 15053371 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070418.3 NP_502819.2 4 - 5 932 74

View Full Product Details