Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srg-51
Entrez Gene ID
189807
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_Y40H7A.8
Chromosome Location
Chr.IV: 15139098 - 15140932 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070426.2 NP_502827.1 1 - 2 483 172

View Full Product Details