Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-5
Entrez Gene ID
191717
Gene Name
protein-coding
Gene Aliases
CELE_Y73F8A.21
Chromosome Location
Chr.IV: 15400277 - 15447947 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276894.4 NP_001263823.2 5 - 6 558 120
NM_001383253.1 5 - 6 555 120
NR_101983.1 5 - 6 489 120

View Full Product Details