Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-16
Entrez Gene ID
190892
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y105C5B.10
Chromosome Location
Chr.IV: 15931822 - 15933914 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070498.2 NP_502899.2 5 - 6 692 68

View Full Product Details