Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lgc-19
Entrez Gene ID
190895
Gene Name
protein-coding
Gene Aliases
CELE_Y105C5B.16
Chromosome Location
Chr.IV: 15956571 - 15962712 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070504.2 NP_502905.2 4 - 5 415 137

View Full Product Details