Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-74
Entrez Gene ID
186932
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_K03D3.1
Chromosome Location
Chr.IV: 16329459 - 16331059 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070544.1 NP_502945.1 1 - 2 159 150

View Full Product Details