Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rom-3
Entrez Gene ID
191000
Gene Name
protein-coding
Gene Aliases
CELE_Y116A8C.14
Chromosome Location
Chr.IV: 16965284 - 16970490 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313037.1 NP_001299966.1 4 - 5 1453 86
NM_070612.2 NP_503013.2 12 - 13 2410 86

View Full Product Details