Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
atgp-1
Entrez Gene ID
178504
Gene Name
protein-coding
Gene Aliases
CELE_F26D10.9
Chromosome Location
Chr.IV: 17269342 - 17272181 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380570.1 NP_001367158.1 1 - 2 136 81
NM_070663.7 NP_503064.2 8 - 9 1445 81

View Full Product Details