Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snf-9
Entrez Gene ID
178512
Gene Name
Transporter
Gene Aliases
CELE_C49C3.1
Chromosome Location
Chr.IV: 17312141 - 17315462 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070676.4 NP_503077.2 2 - 3 547 80

View Full Product Details