Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lact-6
Entrez Gene ID
182288
Gene Name
Beta-lactamase domain-containing protein
Gene Aliases
CELE_C06A12.5
Chromosome Location
Chr.IV: 17429597 - 17431387 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070708.6 NP_503109.1 4 - 5 885 136

View Full Product Details