Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cgp-1
Entrez Gene ID
178540
Gene Name
protein-coding
Gene Aliases
CELE_Y38C9A.2
Chromosome Location
Chr.V: 118202 - 122789 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313303.3 NP_001300232.1 2 - 3 282 83
NM_001313304.3 NP_001300233.1 1 - 2 165 83
NM_070736.8 NP_503137.1 5 - 6 1564 83

View Full Product Details