Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-11
Entrez Gene ID
177419
Gene Name
DNaJ domain (prokaryotic heat shock protein)
Gene Aliases
CELE_F38A5.13
Chromosome Location
Chr.IV: 6585871 - 6588186 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068605.7 NP_501006.1 1 - 2 87 60

View Full Product Details