Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
drh-1
Entrez Gene ID
177425
Gene Name
protein-coding
Gene Aliases
CELE_F15B10.2
Chromosome Location
Chr.IV: 6607376 - 6613353 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380283.2 NP_001368373.1 9 - 10 1230 92
NM_068617.8 NP_501018.1 14 - 15 2009 92
GenBank mRNA AF480439.1 - 14 - 15 2004 92

View Full Product Details