Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dyci-1
Entrez Gene ID
177440
Gene Name
protein-coding
Gene Aliases
CELE_C17H12.1
Chromosome Location
Chr.IV: 6779774 - 6785272 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383110.1 3 - 4 492 100

View Full Product Details