Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srh-78
Entrez Gene ID
191872
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_DC2.2
Chromosome Location
Chr.V: 217147 - 219513 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070755.2 NP_503156.1 2 - 3 362 105

View Full Product Details