Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-39
Entrez Gene ID
178553
Gene Name
protein-coding
Gene Aliases
CELE_R02C2.3
Chromosome Location
Chr.V: 246608 - 251756 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313231.3 NP_001300160.1 2 - 3 390 73
NM_001313232.3 NP_001300161.1 2 - 3 255 73

View Full Product Details