Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atg-9
Entrez Gene ID
178561
Gene Name
protein-coding
Gene Aliases
CELE_T22H9.2
Chromosome Location
Chr.V: 349385 - 359658 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377584.1 NP_001364810.1 1 - 2 51 60
NM_070777.5 NP_503178.2 10 - 11 2400 60

View Full Product Details