Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-35
Entrez Gene ID
191854
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_M02H5.11
Chromosome Location
Chr.V: 447531 - 449230 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070804.2 NP_503205.1 4 - 5 765 115

View Full Product Details