Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lipl-3
Entrez Gene ID
178572
Gene Name
Lipase lipl-3
Gene Aliases
CELE_R11G11.14
Chromosome Location
Chr.V: 521595 - 524218 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070832.4 NP_503233.1 6 - 7 1109 79

View Full Product Details