Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-90
Entrez Gene ID
178577
Gene Name
Nuclear hormone receptor family member nhr-90
Gene Aliases
CELE_ZK488.2
Chromosome Location
Chr.V: 610445 - 612513 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070862.5 NP_503263.1 2 - 3 334 71

View Full Product Details