Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-17
Entrez Gene ID
185994
Gene Name
protein-coding
Gene Aliases
CELE_F48G7.6
Chromosome Location
Chr.V: 623298 - 625022 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313180.1 NP_001300109.1 2 - 3 608 147
NM_001313183.1 NP_001300112.1 1 - 2 359 147

View Full Product Details