Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-24
Entrez Gene ID
185171
Gene Name
protein-coding
Gene Aliases
CELE_F31F4.13
Chromosome Location
Chr.V: 667896 - 671442 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313161.1 NP_001300090.1 3 - 4 600 134
NM_070886.2 NP_503287.2 3 - 4 600 134
NR_132481.1 3 - 4 600 134

View Full Product Details