Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-4
Entrez Gene ID
190064
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y49C4A.5
Chromosome Location
Chr.V: 690875 - 692770 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070895.1 NP_503296.1 2 - 3 329 130

View Full Product Details